2',3',4'-Trifluoroacetophenone ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 1-(2,3,4-Trifluorophenyl)ethanone, 1-(2,3,4-Trifluorophenyl)ethan-1-one

T3425-1G
TCIAT3425-1GEA 51.7 EUR
TCIAT3425-1G
2',3',4'-Trifluoroacetophenone ≥98.0% (by GC)
2',3',4'-Trifluoroacetophenone
Formule: C₈H₅F₃O
MDL Nummer: MFCD00061231
CAS nummer: 243448-15-9

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR