Tissue-clearing reagent, TOMEI for plants

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

T3530-100ML
TCIAT3530-100MLEA 84.7 EUR
TCIAT3530-100ML
Tissue-clearing reagent, TOMEI for plants
Kleurstoffen Microscopy and staining reagents

Specifications
Appearance: Colorless to Light yellow clear liquid
Functionality test (transparency) (display): to pass test

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR