Trihexyl(tetradecyl)phosphonium dicyanamide ≥93.0% (by total nitrogen basis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

T3534-5G T3534-25G
TCIAT3534-5GEA 36.3 EUR
TCIAT3534-5G TCIAT3534-25G
Trihexyl(tetradecyl)phosphonium dicyanamide ≥93.0% (by total nitrogen basis)
Trihexyl(tetradecyl)phosphonium dicyanamide
Formule: C₃₄H₆₈N₃P
MDL Nummer: MFCD04117699
CAS nummer: 701921-71-3
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=696514bfb7f4903e70f81c68943efc89139f635b&vid=696514bfb7f4903e70f81c68943efc89139f635b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Brown clear liquid
Purity (with Total Nitrogen) min. 93.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR