Tetrabutylammonium nitrate ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

T3651-25G T3651-5G
TCIAT3651-25GEA 86.9 EUR
TCIAT3651-25G TCIAT3651-5G
Tetrabutylammonium nitrate ≥98.0% (by titrimetric analysis)
Tetrabutylammonium nitrate
Formule: C₁₆H₃₆N₂O₃
MW: 304.47 g/mol
MDL Nummer: MFCD00043202
CAS nummer: 1941-27-1
EINECS: 217-726-0
UN: 1479
ADR: 5.1 II

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 117.0 to 121.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR