Tissue-clearing reagent, CUBIC-B for animals

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

T3780-100ML T3780-25ML
TCIAT3780-100MLEA 143 EUR
TCIAT3780-100ML TCIAT3780-25ML
Tissue-clearing reagent, CUBIC-B for animals
Kleurstoffen Microscopy and staining reagents

Specifications
Appearance: Colorless to Almost colorless clear liquid
Functionality test: to pass test (Remaining fluorescence strength)
Decalcification test: max. 0.3 (590 to 600 nm)

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR