1,3,5-Undecatriene (mixture of isomers) ≥75.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

U0062-5ML U0062-25ML
TCIAU0062-5MLEA 69.3 EUR
TCIAU0062-5ML TCIAU0062-25ML
1,3,5-Undecatriene (mixture of isomers) ≥75.0% (by GC)
1,3,5-Undecatriene (mixture of isomers)
Formule: C₁₁H₁₈
Vlampunt: 65°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00015699
CAS nummer: 16356-11-9
EINECS: 240-416-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1f02b7772daecb4a503acf1e466490c08cb70a8b&vid=1f02b7772daecb4a503acf1e466490c08cb70a8b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 75.0% (total of isomer)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR