Undecafluoropentyl Iodide ≥98.0% (by GC) stabilised

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing


Stabilisatie: Stabilised with copper chip

U0081-25G U0081-5G
TCIAU0081-25GEA 384 EUR
TCIAU0081-25G TCIAU0081-5G
Undecafluoropentyl Iodide ≥98.0% (by GC) stabilised
Undecafluoropentyl Iodide
Formule: C₅F₁₁I
Kookpunt: 94°C
Smeltpunt: –50°C
MDL Nummer: MFCD08461629
CAS nummer: 638-79-9
EINECS: 211-350-0

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR