Sulphisoxazole ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

U0098-25G
TCIAU0098-25GEA 197 EUR
TCIAU0098-25G
Sulphisoxazole ≥98.0% (by HPLC)
Sulphisoxazole
Formule: C₁₁H₁₃N₃O₃S
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00003150
CAS nummer: 127-69-5
EINECS: 204-858-9
Merck Index: 14,08952

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 193.0 to 197.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR