1-Vinyl-2-pyrrolidone ≥99.0% (by GC) stabilised

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar


Stabilisatie: Stabilised with N,N'-di-sec-butyl-p-phenylenediamine

V0026-25ML V0026-500ML
TCIAV0026-25MLEA 20.9 EUR
TCIAV0026-25ML TCIAV0026-500ML
1-Vinyl-2-pyrrolidone ≥99.0% (by GC) stabilised
1-Vinyl-2-pyrrolidone
Formule: C₆H₉NO
Kookpunt: 148°C
Vlampunt: 102°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00003197
CAS nummer: 88-12-0
EINECS: 201-800-4
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=33698aaa9705f88f04b37a310a220a4d5cc75060&vid=33698aaa9705f88f04b37a310a220a4d5cc75060&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Whiteor Colorless to Light yellow to Red powder to lump to clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR