Valeraldehyde 2,4-Dinitrophenylhydrazone ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Pentanal 2,4-Dinitrophenylhydrazone

V0066-1G V0066-10G
TCIAV0066-1GEA 24.2 EUR
TCIAV0066-1G TCIAV0066-10G
Valeraldehyde 2,4-Dinitrophenylhydrazone ≥98.0% (by HPLC, titration analysis)
Valeraldehyde 2,4-Dinitrophenylhydrazone
Formule: C₁₁H₁₄N₄O₄
MW: 266.26 g/mol
MDL Nummer: MFCD00191595
CAS nummer: 2057-84-3
UN: 1325
ADR: 4.1 III

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 108.0 to 111.0 deg-C

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR