Zinc (II) phthalocyanine (Sulphate ionophore II) ≥98.0% (by titrimetric analysis), purified by sublimation

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Z0037-500MG
TCIAZ0037-500MGEA 251 EUR
TCIAZ0037-500MG
Zinc (II) phthalocyanine (Sulphate ionophore II) ≥98.0% (by titrimetric analysis), purified by sublimation
Zinc (II) phthalocyanine (Sulphate ionophore II)
Formule: C₃₂H₁₆N₈Zn
MDL Nummer: MFCD00041955
CAS nummer: 14320-04-8
EINECS: 238-262-5

Order Now

Specification Test Results

Appearance Dark red to Dark purple to Dark blue powder to crystal
Purity (Chelometric Titration) min. 98.0 %
Molar extinction coefficient min. 225000 (DMF, 667 to 670nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR