PCRmax® Alpha Cyclers
Leverancier: Cole-Parmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
93945-00 93945-04 93945-06 93945-08 93945-10 93945-12 93945-20 93945-22 93945-28
TECN93945-00EA 5008.1 EUR
TECN93945-00 TECN93945-04 TECN93945-06 TECN93945-08 TECN93945-10 TECN93945-12 TECN93945-20 TECN93945-22 TECN93945-28
PCRmax® Alpha Cyclers
PCR-toestellen
Easily create, modify and access programs, saving time.
  • Flexible—your choice of one, two, or four block capacities in 96-well, 384-well, or mixed formats
  • Independently control each block for simplified temperature and space optimization
  • Software allows quick access to commonly used protocols
These compact thermal cyclers deliver reproducible results every run—with little to no training needed. Features include a clear responsive touch screen, secure user-specific programming, adjustable heated lid, and active sample cooling for sharper amplification.
More Product Information