Polished mirrors
Leverancier: TEKNOMEK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
CRMI-400X900FH CRMI-PMFH18X60 CRMI-600X400FH CRMI-600X600 CRMI-R450FH CRMI-300X300 CRMI-300X600
TEKMCRMI-400X900FHEA 0 EUR
TEKMCRMI-400X900FH TEKMCRMI-PMFH18X60 TEKMCRMI-600X400FH TEKMCRMI-600X600 TEKMCRMI-R450FH TEKMCRMI-300X300 TEKMCRMI-300X600
Polished mirrors
Spiegels
Our exceptionally hygienic mirrors are built to handle the toughest of environments.
  • Shatterproof and safe
  • De-burred edges and rounded corners to prevent injury
  • 1,2 mm thick stainless steel
  • Choose with or without pre-drilled mounting holes
  • Vandal resistant
  • Material: 304-grade stainless steel
Not only exceptionally hygienic, but also durable and long-lasting, our steel mirrors are made from 304-grade, meaning they can handle even the toughest of environments without breaking or shattering.
They have de-burred edges and rounded corners to preventMore Product Information