Enclosed PPE dispensers
Leverancier: TEKNOMEK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
HWDI-MG4S DPPE-G4F DPPE-GF HWDI-MG1S HWDI-MG4
TEKMHWDI-MG4SEA 0 EUR
TEKMHWDI-MG4S 112-6573 112-6571 TEKMHWDI-MG1S TEKMHWDI-MG4
Enclosed PPE dispensers
PBM-dispensers
Our enclosed PPE dispenser offers intelligent design for all your PPE needs with an easy-clean sloping top.
  • Easy to clean intelligent design
  • Sloping top
  • Design has no plastic parts
  • Available in 316-grade stainless steel
  • Brushed dull polish finish
  • With or without flaps
  • Wall fixed - option to add magnets for no damage wall fixing
Our glove box and face mask dispenser is intelligently designed as it comes with no plastic parts and with a sloping top that makes it very easy to clean.
The glove box/face mask dispenser is manufactured in 304-gradeMore Product Information