Cryoboxes, Cardboard
Leverancier: TENAK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
TE22871 TE22872 TE22874 TE22875 TE22876 TE22878 TE22879 TE22870 TE22880
TENATE22871EA 82.4 EUR
TENATE22871 TENATE22872 TENATE22874 TENATE22875 TENATE22876 TENATE22878 TENATE22879 TENATE22870 TENATE22880
Cryoboxes, Cardboard
Dozen Dozen voor cryobuisjes
Colored cryoboxes possible to orient in different ways for classic colour coding, tracking work in progress, identifying the owner of the box, and monitoring sample thawing.
  • PP coated water-repellent surface
  • Internal printed alpha numeric grid
For space-saving solutions, the cryobox is also available in a convenient flat-packed Origami box format.
Order Now
Specificaties
Environmentally Preferable