Microplate racks for chest freezers
Leverancier: TENAK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
TE27114 TE27178 TE27100 TE27102 TE27104 TE27106 TE27108 TE27110 TE27112 TE27116 TE27130 TE27132 TE27134 TE27136 TE27138 TE27140 TE27142 TE27144 TE27146 TE27160 TE27162 TE27164 TE27166 TE27168 TE27170 TE27172 TE27174 TE27176 TE27180 TE27190 TE27192 TE27194 TE27196 TE27198 TE27200 TE27202 TE27204 TE27210 TE27212 TE27214 TE27216 TE27218 TE27220
TENATE27114EA 202.35 EUR
TENATE27114 TENATE27178 TENATE27100 TENATE27102 479-2178 TENATE27106 TENATE27108 TENATE27110 TENATE27112 TENATE27116 TENATE27130 TENATE27132 479-2179 TENATE27136 TENATE27138 TENATE27140 TENATE27142 TENATE27144 TENATE27146 TENATE27160 TENATE27162 479-2180 TENATE27166 TENATE27168 TENATE27170 TENATE27172 TENATE27174 TENATE27176 TENATE27180 TENATE27190 TENATE27192 479-2181 TENATE27196 TENATE27198 TENATE27200 TENATE27202 TENATE27204 TENATE27210 479-2182 TENATE27214 TENATE27216 TENATE27218 TENATE27220
Microplate racks for chest freezers
Rekken Rekken voor microplaten
Stainless steel standard microplate racks.
  • With a locking rod to secure cryoboxes