Comfort microplate racks for upright freezers
Leverancier: TENAK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
TE35400 TE35402 TE35404 TE35406 TE35410 TE35412 TE35414 TE35416 TE35420 TE35422 TE35424 TE35426 TE35430 TE35432 TE35434 TE35440 TE35442 TE35450 TE35452 TE35454 TE35456 TE35458 TE35460 TE35462 TE35464 TE35466 TE35468 TE35470 TE35480 TE35482 TE35484 TE35486 TE35488 TE35490 TE35500 TE35502 TE35504 TE35506 TE35508 TE35510 TE35512 TE35514 TE35520 TE35522 TE35524 TE35526 TE35530 TE35532 TE35534 TE35536 TE35540 TE35542 TE35544 TE35546 TE35548 TE35550 TE35552 TE35554 TE35556 TE35558 TE35560 TE35562 TE35564 TE35570 TE35574 TE35576 TE35580 TE35582 TE35584 TE35586 TE35590 TE35592 TE35594 TE35596 TE35598 TE35600 TE35602 TE35604 TE35606 TE35610 TE35612 TE35614
TENATE35400EA 342.71 EUR
TENATE35400 TENATE35402 TENATE35404 TENATE35406 TENATE35410 TENATE35412 TENATE35414 TENATE35416 TENATE35420 TENATE35422 TENATE35424 TENATE35426 TENATE35430 TENATE35432 TENATE35434 TENATE35440 TENATE35442 TENATE35450 TENATE35452 479-2191 TENATE35456 TENATE35458 TENATE35460 TENATE35462 479-2192 TENATE35466 TENATE35468 TENATE35470 TENATE35480 TENATE35482 TENATE35484 479-2193 TENATE35488 TENATE35490 TENATE35500 TENATE35502 479-2194 TENATE35506 TENATE35508 TENATE35510 479-2195 TENATE35514 TENATE35520 TENATE35522 479-2130 TENATE35526 TENATE35530 TENATE35532 479-2131 TENATE35536 TENATE35540 TENATE35542 TENATE35544 479-2132 TENATE35548 TENATE35550 TENATE35552 479-2133 TENATE35556 TENATE35558 TENATE35560 479-2134 TENATE35564 TENATE35570 TENATE35574 TENATE35576 TENATE35580 TENATE35582 TENATE35584 TENATE35586 TENATE35590 TENATE35592 TENATE35594 TENATE35596 TENATE35598 TENATE35600 TENATE35602 TENATE35604 TENATE35606 TENATE35610 TENATE35612 TENATE35614
Comfort microplate racks for upright freezers
Rekken Rekken voor microplaten
This is a complete range of racks for microtiterplates for use in chest freezers.
Choose from 5 different shelf heights depending on preferences. The racks can either be used as one plate per shelf or the plates can be stacked. The racks are available with a shelf height of 16 mm, 25 mm, 45 mm, 65 mm and 85 mm. The external footprint is 135×92 mm on all racks. The comfort rack has an alert preventing the shelves from sliding out.
Order Now
Specificaties
Environmentally Preferable