Intraveneuze katheter, Surflash™
Leverancier: TERUMO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SR-FF1464 SR-FF1651 SR-FF1832 SR-FF1851 SR-FF2032 SR-FF2051 SR-FF2225 SR-FF2419
TERUSR-FF1464EA 122 EUR
TERUSR-FF1464 TERUSR-FF1651 TERUSR-FF1832 TERUSR-FF1851 TERUSR-FF2032 TERUSR-FF2051 TERUSR-FF2225 TERUSR-FF2419
Intraveneuze katheter, Surflash™
Canules
Surflash is made of PU, which is firm at room temperature but soft at body temperature, making the catheter more comfortable.
  • Only exists in the straight version
  • Enables quick and easy confirmation of the placement in the vein by an additional flashback
  • Higher certainty of proper placement
Helpful with patients whose veins are hard to detect and in which venipuncture is more difficult, such as small children, people undergoing chemotherapy.