Differential thermometer for TC type K with app connection, testo 922
Leverancier: Testo
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0563.0922
TEST0563.0922EA 157 EUR
TEST0563.0922
Differential thermometer for TC type K with app connection, testo 922
Thermometers Digitale thermometers
Compact testo 922 is versatile instrument: it not only determines temperatures quickly and accurately, but also does a direct calculation of the differential temperature. This means that measurements can be completed just as quickly as documentation with the practical testo Smart App for smartphones and tablets. A typical application for the testo 922 is the control of the supply/return temperature on heating circuit manifolds. Two Type K thermocouple probes are included, but the testo 922 is also compatible with other commercially available TC type K probes.
  • Wide range of applications