Luxmeter, testo 545
Leverancier: Testo
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0563.1545
TEST0563.1545EA 406 EUR
TEST0563.1545
Luxmeter, testo 545
Lichtmeters
Lux meter testo 545 can measure the illuminance of all common light sources quickly, easily and precisely to ensure productivity, performance and health in workplaces. By automatically calculating the time and point average value, the Lux meter shows all relevant information at a glance.
  • Measurement of illuminance (Lux) according to the V-lambda curve for all common light sources
  • Compatible with almost all commercially available LEDs and thus ensures a wide range of applications (except single-colour blue LED)
  • Audible alarm sounds if a limit value is
More Product Information