Air flow comboKit 1 with Bluetooth®, Testo 440
Leverancier: Testo
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0563.4406
TEST0563.4406EA 969 EUR
TEST0563.4406
Air flow comboKit 1 with Bluetooth®, Testo 440
Dataloggers
Airflow Combokit 1 equipped with clearly structured measurement menu for volume flow and parallel determination of air velocity, differential pressure, humidity and temperature in ventilation ducts or at ventilation outlets.
  • Long-term monitoring of indoor air quality
  • Combine the vane probe with Bluetooth with the telescope for ceiling outlets
  • Turbulence measurement in accordance with EN ISO 7730 / ASHRAE 55
Convenient for wireless probe, internal data storage and USB port for data export. Ideal for long-term monitoring of indoor air quality andMore Product Information
Order Now
Specificaties
Environmentally Preferable
Interface Bluetooth