Wireless monitoring systems, Smart-Vue
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SV206-101 SV104-502 SV200-101 SV200-201 SV201-101 SV201-201 SV202-101 SV202-201 SV203-101 SV203-201 SV204-201 SV100-501 SV209-101 SV509-500 SV500-501 SV212-501 SV205-101 SV209-201 SV208-101 SV207-201 SV207-101 SV206-201 SV103-503 SV106-503-LSB SV103-511-LSB SV103-512-LSB SV103-513-LSB SV103-514-LSB SV603-500-LSB SV604-500-LSB SV605-500-LSB SV606-500-LSB SV607-500-LSB SV608-500-LSB SV609-500-LSB SV610-500-LSB SV611-500-LSB SV612-500-LSB
620-1895EA 515 EUR
620-1895 620-1879 620-1890 THBRSV200-201 620-1891 THBRSV201-201 620-1892 THBRSV202-201 620-1893 THBRSV203-201 THBRSV204-201 620-1887 THBRSV209-101 620-1898 620-1897 THBRSV212-501 620-1894 THBRSV209-201 620-1896 THBRSV207-201 620-1876 THBRSV206-201 620-1877 THBRSV106-503-LSB THBRSV103-511-LSB THBRSV103-512-LSB THBRSV103-513-LSB THBRSV103-514-LSB THBRSV603-500-LSB THBRSV604-500-LSB THBRSV605-500-LSB THBRSV606-500-LSB THBRSV607-500-LSB THBRSV608-500-LSB THBRSV609-500-LSB THBRSV610-500-LSB THBRSV611-500-LSB THBRSV612-500-LSB
Wireless monitoring systems, Smart-Vue
Bewakingssystemen Bewakingssystemen voor CO2 en temperatuur
Ambient measurements or monitoring of laboratory equipment.
  • Easy to read display of monitored setpoints directly on module
  • Wireless data transmission (868 MHz): No wiring or rebuilding required for installation
  • Fully integrated open software: Data can easily be exported to Excel, Word or PDF
  • Safety signal transfer: Automatic signal recognition makes it possible to use each module as repeater
  • Accessible anywhere via the internet with Smart-Vue client software
These instruments safeguard the integrity of precious samples by continuously monitoring andMore Product Information
Order Now
Specificaties
Environmentally Preferable