Labelling tapes for anesthesia drug syringes
Leverancier: PRECISION DYNAMICS TIMEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
AN-21 AN-64 AN-105 AN-4 AN-150 AN-20 AN-7 AN-13 AN-120 AN-47 AN-5 AN-134 AN-10 AN-41 AN-1 ANS-84 AN-60 AN-76 AN-147 ANS-71 AN-101 ANS-16 AN-35 AN-95 AN-122 AN-106 AN-51 AN-77 AN-31 AN-113 AN-79 AN-39 AN-59 AN-62 AN-11 AN-57 AN-40 AN-78 AN-3 AN-19 AN-68 AN-42 AN-109 AN-54 AN-2 AN-18 AN-103 ANS-50 ANS-15 AN-12 ANS-14 AN-6 AN-72 AN-75 AN-81 ANS-17 AN-110 AN-132 AN-155 AN-89 AN-90
TIMEAN-21EA 24.9 EUR
TIMEAN-21 TIMEAN-64 TIMEAN-105 TIMEAN-4 TIMEAN-150 TIMEAN-20 TIMEAN-7 TIMEAN-13 TIMEAN-120 TIMEAN-47 TIMEAN-5 TIMEAN-134 TIMEAN-10 TIMEAN-41 TIMEAN-1 TIMEANS-84 TIMEAN-60 TIMEAN-76 TIMEAN-147 TIMEANS-71 TIMEAN-101 TIMEANS-16 TIMEAN-35 TIMEAN-95 TIMEAN-122 TIMEAN-106 TIMEAN-51 TIMEAN-77 TIMEAN-31 TIMEAN-113 TIMEAN-79 TIMEAN-39 TIMEAN-59 TIMEAN-62 TIMEAN-11 TIMEAN-57 TIMEAN-40 TIMEAN-78 TIMEAN-3 TIMEAN-19 TIMEAN-68 TIMEAN-42 TIMEAN-109 TIMEAN-54 TIMEAN-2 TIMEAN-18 TIMEAN-103 TIMEANS-50 TIMEANS-15 TIMEAN-12 TIMEANS-14 TIMEAN-6 TIMEAN-72 TIMEAN-75 TIMEAN-81 TIMEANS-17 TIMEAN-110 TIMEAN-132 TIMEAN-155 TIMEAN-89 TIMEAN-90
Labelling tapes for anesthesia drug syringes
Etiketten
Coated paper tape with removable adhesive. The following user applied drug tapes are available for accurate and reliable labelling of all anesthesia drug syringes. This specification covers the size, colour, pattern, and type used on labels applied to unlabelled syringes, filled by the users or their agents, to identify the drug content. In addition, tapes correspond to the published 2004 JCAHO Standard MM,4,30 for appropriately labelling all medications with drug name, strength, amount, expiration date, expiration time, and initials of the person administering the medication.
Order Now
Specificaties
Environmentally Preferable