Magnetic hotplate stirrer, hotMIX 1
Leverancier: 2MAG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
61001_CH 61001_UK
TMAG61001_CHEA 553 EUR
TMAG61001_CH TMAG61001_UK
Magnetic hotplate stirrer, hotMIX 1
Roerders Magnetische roerders met verwarmingsplaat
Compact, powerful heatable single point stirrer suitable for the operation with viscous and aggressive media in chemical, biotechnical and medical applications.
  • Hot plate made from resistant aluminium alloy
  • Price-optimised magnetic stirrer
  • 100% maintenance-free and wear-free
  • Speed range from 100 to 1600 min⁻¹
  • Jerk-free stirring even at low speeds
SoftStart for reliable catching/centering and safe accelerating of the stirring bar, resistant stainless steel housing for easy and fast cleaning, spill over protection, extremely robust construction. StirrerMore Product Information