Size exclusion chromatography sorbents, Toyopearl®
Leverancier: TOSOH Bioscience
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0007448 0007451 0007453 0007455 0007457 0007459 0007465 0007466 0007467 0007469 0014682 0014686 0014687 0014688 0014691 0019810 0019815 0019807 0019808 0019811 0019812 0019816 0019813 0019819 0019820 0019821 0019814 0014683 0007449 0014681 0014684 0014685 0014689 0014690 0021481
TOYO0007448EA 789 EUR
TOYO0007448 TOYO0007451 TOYO0007453 TOYO0007455 TOYO0007457 TOYO0007459 TOYO0007465 TOYO0007466 TOYO0007467 TOYO0007469 TOYO0014682 TOYO0014686 TOYO0014687 TOYO0014688 TOYO0014691 TOYO0019810 TOYO0019815 TOYO0019807 TOYO0019808 TOYO0019811 TOYO0019812 TOYO0019816 TOYO0019813 TOYO0019819 TOYO0019820 TOYO0019821 TOYO0019814 TOYO0014683 TOYO0007449 TOYO0014681 TOYO0014684 TOYO0014685 TOYO0014689 TOYO0014690 TOYO0021481
Size exclusion chromatography sorbents, Toyopearl®
Sorbenten voor chromatografie Sorbenten voor HPLC
Toyopearl resins can be used in organic solvents or mixtures of organic solvents and water. The compatibility of DMF with Toyopearl also permits the separation of hydrophobic substances such as polystyrenes.
  • Removal of surfactants such as Triton® X-100 from biological solutions by an adsorption mechanism
  • Use in hydrophobic interaction chromatography (HIC) for the separation of very hydrophobic molecules
  • Suitable for HIC separations as a guard column for hydrophobic impurities
  • Used as a stationary phase for either normal or reversed phase separations depending on solvent system selected
The Toyopearl HW-type resins are commonly used in size exclusion chromatography and desalting applications.