Anion exchange chromatography columns, TSK-GEL®, sugar
Leverancier: TOSOH Bioscience
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0008639 0008640
TOYO0008639EA 1790 EUR
TOYO0008639 TOYO0008640
Anion exchange chromatography columns, TSK-GEL®, sugar
Kolommen voor chromatografie
TSK-GEL® sugar columns are polymer based stainless steel columns for analysis of monosacchrides, disaccharides and sugar alcohols by isocratic analysis (TSKgel sugar analysis AXI) or by gradient analysis (TSKgel sugar analysis AXG).