Hydrophobic interaction chromatography resins, Toyopearl®, TSK-GEL®
Leverancier: TOSOH Bioscience
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0043150 0043175 0019806 0043125 0043278
TOYO0043150EA 789 EUR
TOYO0043150 TOYO0043175 TOYO0019806 TOYO0043125 TOYO0043278
Hydrophobic interaction chromatography resins, Toyopearl®, TSK-GEL®
Sorbenten voor chromatografie Sorbenten voor HPLC
These are bulk media products which contain three or four different ligand types offered for a chromatography mode.
Physical properties of resins in different buffer systems can be identified by slurry and slurry mechanics, resin handling during column packing, mechanical strength relative to other resin backbones and degree of compressibility.
It is used to determine the resin's dynamic binding capacity, selectivity, column efficiency, operating conditions by developmental scientist or engineer.
Order Now
Specificaties
Environmentally Preferable