Beakers, low form, quartz, with spout
Leverancier: TECHNICAL QUARTZ SOLUTIONS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
75200 75201 75202 75203 75204 75205 75206 75207 75208 75209 75210 75211 75212
TSLA75200EA 24.2 EUR
TSLA75200 TSLA75201 TSLA75202 TSLA75203 TSLA75204 TSLA75205 TSLA75206 TSLA75207 TSLA75208 TSLA75209 TSLA75210 TSLA75211 TSLA75212
Beakers, low form, quartz, with spout
Bekers
Temperature-resistant beakers made from transparent quartz glass.
  • High chemical resistance
  • High resistance against thermal schock
  • Made for tasks up to 1050 °C
  • High purity
  • Without graduations