(±)-1,2-Propanediol ≥99.5% (dry basis), BAKER ANALYZED® ACS, J.T.Baker®

Leverancier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Propylene glycol, 1,2-Propanediol, (±)-1,2-Propylene glycol, 1,2-Dihydroxypropane, (±)-1,2-Dihydroxy propane

U510-07EA 257 EUR
U510-07
(±)-1,2-Propanediol ≥99.5% (dry basis), BAKER ANALYZED® ACS, J.T.Baker®
(±)-1,2-Propanediol
Formule: CH₃CH(OH)CH₂OH
MW: 76.1 g/mol
Kookpunt: 187.6
Smeltpunt: –59 °C
Dichtheid: 1.04 g/cm³ (20 °C)
Vlampunt: 99
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064272
CAS nummer: 57-55-6
EINECS: 200-338-0
Merck Index: 13,07947
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2af195b87767feb8d1baca34925153d6f97ae2d&vid=a598a9042847dc3ce52daed1a9326b592b163170&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements = 99.5 %
For Laboratory,Research,or Manufacturing Use = 99.5 %
ACS - Assay (CH₃CHOHCH₂OH) (corrected for water) ≥99.5 %
ACS - Color (APHA) ≤10
ACS - Residue after Ignition ≤0.005 %
ACS - Titrable Acid (meq/g) ≤0.0005
ACS - Water (H₂O)(by Karl Fischer titrn) ≤0.2 %
Chloride (Cl) ≤1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR