Accessories for face shields
Leverancier: UNIVET
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
605.04.00.00
UNVT605.04.00.00EA 31.6 EUR
UNVT605.04.00.00
Accessories for face shields
Gelaatsschermen
This ergonomic headgear is made from anti-allergic material. With upper band for height adjustment of the cap.
  • Interchangeable front foam sweatband
  • Adjustment of the head circumference by rack
  • 90° flip-up through friction
  • Free of exposed metal parts for protection against electric arcs