Autoclavable cleanroom goggles, 619
Leverancier: UNIVET
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
619.04.23.10 619.05.23.10
UNVT619.04.23.10EA 337 EUR
UNVT619.04.23.10 UNVT619.05.23.10
Autoclavable cleanroom goggles, 619
Ruimzichtbrillen
These cleanroom googles are available with a direct or an indirect ventilation system. Ideal for use in rooms with contamination control (clean and white room) as well as in aseptic environments.
  • Polycarbonate lenses protect against impacts and UV radiations
  • Spherical, panoramic lens: Goggles can be worn over prescription glasses
  • Elastic strap in antistatic silicone has an innovative quick adjustment system
  • Indirect ventilation system in the lower part prevents the escape of particulate matter
  • Can be used for prolonged periods as it does not create
More Product Information