Fittings, Super Flangeless™, Upchurch Scientific®
Leverancier: UPCHURCH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
P-359X P-360 P-360X P-381 P-387 F-156 P-317 P-319 P-331 P-332 P-336 P-350 P-350X P-352 P-357 P-357-2 P-331G
UPCHP-359XEA 76.3 EUR
UPCHP-359X UPCHP-360 UPCHP-360X UPCHP-381 UPCHP-387 UPCHF-156 UPCHP-317 UPCHP-319 UPCHP-331 UPCHP-332 UPCHP-336 UPCHP-350 UPCHP-350X UPCHP-352 UPCHP-357 UPCHP-357-2 UPCHP-331G
Fittings, Super Flangeless™, Upchurch Scientific®
Fittingen en connectoren
These fittings eliminate loosening due to tubing twist.