Tubing, stainless steel, Upchurch Scientific®
Leverancier: UPCHURCH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
TS-632 TS-431 TS-484 TS-706 TS-210 TS-203 TS-171 TS-333 TS-380 TS-541
UPCHTS-632EA 32.7 EUR
UPCHTS-632 UPCHTS-431 UPCHTS-484 UPCHTS-706 UPCHTS-210 UPCHTS-203 UPCHTS-171 UPCHTS-333 UPCHTS-380 UPCHTS-541
Tubing, stainless steel, Upchurch Scientific®
Slangen Chromatography Tubing
Stainless steel tubing.
Seamless, pre-cut 316 stainless steel tubing meets the exacting requirements of today’s analyses. Thorough preparation guarantees that our tubing is truly ready-to-use, with flat-burr-free ends and a clean finish, which ensures zero dead volume connections and good chromatographic results. Each machine-cut end is polished, deburred on the edges and passivated. High-pressure stainless steel tubing can be sterilised by numerous methods; however, the colour-coded band should be removed if present.