HPLC tubing, stainless steel, Upchurch Scientific®
Leverancier: UPCHURCH SCIENTIFIC
Ratings:
Avg. Ratings: 5.0 (1 Review)
Total Ratings: 1
Avg. Ratings: out of 5
U-101 U-102 U-103 U-104 U-105 U-106 U-107 U-108 U-111 U-1113 U-1114 U-1116 U-1117 U-112 U-1120 U-1123 U-1125 U-1128 U-113 U-1130 U-114 U-1140 U-1148 U-115 U-116 U-117 U-118 U-126 U-127 U-128 U-129 U-130 U-131 U-132 U-133 U-134 U-135 U-136 U-137 U-138 U-139 U-140 U-141 U-142 U-143 U-144 U-145 U-146 U-147 U-148 U-149 U-150 U-151 U-152 U-153 U-154 U-155 U-156 U-157 U-158 U-160 U-161 U-162 U-163 U-164 U-165 U-800 U-803 U-825
UPCHU-101EA 11 EUR
UPCHU-101 UPCHU-102 UPCHU-103 UPCHU-104 554-3214 UPCHU-106 554-3215 UPCHU-108 554-3216 554-3217 554-3218 554-3220 554-3221 554-3222 554-3223 554-3226 554-3227 554-3229 UPCHU-113 UPCHU-1130 554-3230 554-3231 554-3238 UPCHU-115 UPCHU-116 UPCHU-117 UPCHU-118 UPCHU-126 UPCHU-127 UPCHU-128 UPCHU-129 UPCHU-130 UPCHU-131 UPCHU-132 UPCHU-133 UPCHU-134 UPCHU-135 UPCHU-136 UPCHU-137 UPCHU-138 UPCHU-139 UPCHU-140 UPCHU-141 UPCHU-142 UPCHU-143 UPCHU-144 UPCHU-145 UPCHU-146 UPCHU-147 UPCHU-148 UPCHU-149 UPCHU-150 UPCHU-151 554-3239 554-3240 554-3241 UPCHU-155 UPCHU-156 UPCHU-157 UPCHU-158 UPCHU-160 UPCHU-161 UPCHU-162 UPCHU-163 UPCHU-164 UPCHU-165 UPCHU-800 UPCHU-803 554-3271
HPLC tubing, stainless steel, Upchurch Scientific®
Slangen Chromatography Tubing
Stainless steel.
  • Seamless and pre-cut to ensure burr-free, flat connections
  • Rockwell hardness (B) is maximum of 95
  • Many sizes feature a colour-coded band for easy ID identification
  • These products are RoHS compliant and meet ASTM A-269-10 and A-213
Each machine-cut end is polished, deburred on the inside and outside edges, and then the entire tubing is passivated. Finally, reagent-grade isopropanol is flushed through each piece. The thorough preparation and cleaning procedure guarantees tubing that is truly ready-to-use, with flat, burr-free ends and aMore Product Information