Drip pans
Leverancier: USBECK Laborgeräte
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
4781 4784
USBE4781EA 150 EUR
USBE4781 USBE4784
Drip pans
Reservoirs/Schalen/Potten Trays/pannen voor gemorste stoffen
Made of stainless steel.
  • Very good chemical resistance
  • Very good temperature resistance
  • Easy to clean
With removable gridiron.