Anti-ACPL2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverancier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

031552-FITC
USBI031552-FITCEA 884 EUR
USBI031552-FITC
Anti-ACPL2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antilichamen
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: ACPL2 (acid phosphatase-like 2)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR