Anti-AXL Rabbit Polyclonal Antibody (Biotin)

Leverancier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

032344-BIOTIN
USBI032344-BIOTINEA 884 EUR
USBI032344-BIOTIN
Anti-AXL Rabbit Polyclonal Antibody (Biotin)
Antilichamen
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50

Type: Primary
Antigen: AXL (AXL receptor tyrosine kinase)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Specificaties

antigeen symbool AXL
Environmentally Preferable
Type antilichaam Primair,AXL receptor tyrosine kinase
Type antilichaam Primair,AXL receptor tyrosine kinase
Clonality Polyklonaal
Conjugatie Biotine
Reactivity Human
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Flowcytometrie Yes
Epitoop N-terminal
Aminozuurgetal 14 to 44
Formulier liquid
Purification purified by Protein G chromatography
Opslagbuffer PBS, pH 7,2
Opslagtemperatuur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR