Anti-CADM2 Rabbit Polyclonal Antibody

Leverancier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

033141
USBI033141EA 667 EUR
USBI033141
Anti-CADM2 Rabbit Polyclonal Antibody
Antilichamen
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: CADM2 (cell adhesion molecule 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Specificaties

antigeen symbool CADM2
Type antilichaam Primair,Cell adhesion molecule 2
Environmentally Preferable
Type antilichaam Primair,Cell adhesion molecule 2
Clonality Polyklonaal
Conjugatie Onconjugeerd
Reactivity Human
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Epitoop N-terminal
Aminozuurgetal 49 to 76
Formulier liquid
Purification purified by Protein A chromatography
Opslagbuffer PBS, pH 7,2, 0,09% sodium azide.
Opslagtemperatuur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR