Anti-FFAR4 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Leverancier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

036217-AP
USBI036217-APEA 884 EUR
USBI036217-AP
Anti-FFAR4 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antilichamen
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: FFAR4 (free fatty acid receptor 4)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Specificaties

antigeen symbool FFAR4
Environmentally Preferable
Type antilichaam Primair,Free fatty acid receptor 4
Type antilichaam Primair,Free fatty acid receptor 4
Clonality Polyklonaal
Conjugatie AP (Alkaline Phosphatase)
Reactivity Human
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Epitoop internal
Aminozuurgetal 230 to 260
Formulier liquid
Purification purified by Protein A chromatography
Opslagbuffer PBS, pH 7,2, contains ~0,09% sodium azide
Opslagtemperatuur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR