Anti-TBCA Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Leverancier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

042644-PE
USBI042644-PEEA 884 EUR
USBI042644-PE
Anti-TBCA Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antilichamen
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: TBCA (tubulin folding cofactor A)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Specificaties

antigeen symbool TBCA
Environmentally Preferable
Type antilichaam Primair,tubulin folding cofactor A
Type antilichaam Primair,tubulin folding cofactor A
Clonality Polyklonaal
Conjugatie PE (fycoerythrin)
Reactivity Human
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Epitoop N-terminal
Aminozuurgetal 14 to 44
Formulier liquid
Purification purified by Protein A chromatography
Opslagbuffer PBS, pH 7,2
Opslagtemperatuur Cool
Shipping temperature Ice
Tested applications Fluoresence ELISA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR