Anti-TTLL8 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Leverancier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

043442-APC
USBI043442-APCEA 884 EUR
USBI043442-APC
Anti-TTLL8 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antilichamen
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: TTLL8 (tubulin tyrosine ligase-like family, member 8)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Specificaties

antigeen symbool TTLL8
Type antilichaam Primair,tubulin tyrosine ligase-like family, member 8
Environmentally Preferable
Type antilichaam Primair,tubulin tyrosine ligase-like family, member 8
Clonality Polyklonaal
Conjugatie APC (Allophycocyanin)
Reactivity Human
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Epitoop N-terminal
Aminozuurgetal 37 to 67
Formulier liquid
Purification purified by Protein A chromatography
Opslagbuffer PBS, pH 7,2, contains ~0,09% sodium azide
Opslagtemperatuur Cool
Shipping temperature Ice
Tested applications Fluoresence ELISA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR