Anti-DCK Rabbit Polyclonal Antibody

Leverancier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

125666
USBI125666EA 696 EUR
USBI125666
Anti-DCK Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: DCK (deoxycytidine kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Specificaties

antigeen symbool DCK
Environmentally Preferable
Type antilichaam Primair,deoxycytidine kinase
Type antilichaam Primair,deoxycytidine kinase
Clonality Polyklonaal
Conjugatie Onconjugeerd
Reactivity Human
Host Rabbit
Isotype IgG
Western blot Yes
Aminozuurgetal 1 to 260
Formulier liquid
Purification purified by Protein A chromatography
Opslagbuffer PBS, pH 7,2.
Opslagtemperatuur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR