Anti-Glucose Regulated Protein 94 Rabbit Polyclonal Antibody

Leverancier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

136781
USBI136781EA 366 EUR
USBI136781
Anti-Glucose Regulated Protein 94 Rabbit Polyclonal Antibody
Antilichamen
Recommended dilution: Western Blot: 1:10000; Immunoprecipitation: 1:80

Type: Primary
Antigen: Glucose Regulated Protein 94
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR