Anti-Protein Gene Product 9.5 Mouse Monoclonal Antibody [clone: 31A3]

Leverancier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

138591-100
USBI138591-100EA 654 EUR
USBI138591-100
Anti-Protein Gene Product 9.5 Mouse Monoclonal Antibody [clone: 31A3]
Antilichamen
Recommended dilution: Western Blot: 0.5-1 ?g/ml; Immunochemistry: 0.5-1 ?g/ml

Type: Primary
Antigen: Protein Gene Product 9.5
Clonality: monoclonal
Clone: 31A3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Bovine, Human, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR