Anti-Glutamate Decarboxylase 65 Mouse Monoclonal Antibody [clone: 13j39]

Leverancier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

140461
USBI140461EA 543 EUR
USBI140461
Anti-Glutamate Decarboxylase 65 Mouse Monoclonal Antibody [clone: 13j39]
Antilichamen
Recommended dilution: ELISA: 1:100-200; Western Blot: 1:100-400; Immunochemistry: 1:100-500

Type: Primary
Antigen: Glutamate Decarboxylase 65
Clonality: monoclonal
Clone: 13j39
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Rat
Order Now

Specificaties

antigeen symbool GAD65
Type antilichaam Glutamate decarboxylase 65,Primair
Environmentally Preferable
Type antilichaam Glutamate decarboxylase 65,Primair
Clonality Monoklonaal
Conjugatie Onconjugeerd
Clone 13j39
Reactivity Rat
Host Mouse
Isotype
Western blot Yes
ELISA Yes
ImmunoChemie Yes
Formulier liquid
Purification purified by immunoaffinity chromatography
Concentratie ~0,5 mg/ml
Opslagbuffer PBS, pH 7,2, 0,05% sodium azide, 50% glycerol.
Opslagtemperatuur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR