Anti-DKK1 Goat Polyclonal Antibody

Leverancier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

150034
USBI150034EA 809 EUR
USBI150034
Anti-DKK1 Goat Polyclonal Antibody
Antilichamen
Recommended dilution: ELISA: 1:128000; Western Blot: 1 ?g/ml

Type: Primary
Antigen: DKK1 (dickkopf WNT signaling pathway inhibitor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Specificaties

antigeen symbool DKK1
Environmentally Preferable
Type antilichaam Primair,dickkopf WNT signaling pathway inhibitor 1
Type antilichaam Primair,dickkopf WNT signaling pathway inhibitor 1
Clonality Polyklonaal
Conjugatie Onconjugeerd
Reactivity Human
Host Goat
Isotype
Western blot Yes
ELISA Yes
Formulier liquid
Purification purified by immunoaffinity chromatography
Concentratie ~0,5 mg/ml
Opslagbuffer Tris saline, pH 7,3, 0,5% BSA, 0,02% sodium azide.
Opslagtemperatuur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR