Anti-HCST Goat Polyclonal Antibody

Leverancier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

150302
USBI150302EA 688 EUR
USBI150302
Anti-HCST Goat Polyclonal Antibody
Antilichamen
Recommended dilution: ELISA: 1:128000; Western Blot: 0.2-0. 6?g/ml

Type: Primary
Antigen: HCST (hematopoietic cell signal transducer)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR