Anti-MLLT10 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Leverancier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A0923-60C-APC
USBIA0923-60C-APCEA 884 EUR
USBIA0923-60C-APC
Anti-MLLT10 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antilichamen
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:10-1:50
Do not freeze!

Type: Primary
Antigen: MLLT10 (myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 10)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR