Anti-AIRE Rabbit Polyclonal Antibody

Leverancier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A1059-88L
USBIA1059-88LEA 466 EUR
USBIA1059-88L
Anti-AIRE Rabbit Polyclonal Antibody
Antilichamen
Recommended dilution: Western Blot: 4-6?g/ml

Type: Primary
Antigen: AIRE (autoimmune regulator)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR