Anti-Bone Morphogenetic Protein Receptor Type 1B Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 11C3]

Leverancier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

B2553-63F
USBIB2553-63FEA 843 EUR
USBIB2553-63F
Anti-Bone Morphogenetic Protein Receptor Type 1B Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 11C3]
Antilichamen
Do not freeze! Stable at 4°C for 12 months after receipt as an undiluted liquid Dilute required amount only prior to immediate use Further dilutions can be made in assay buffer Caution: APC conjugates are sensitive to light For maximum recovery of product, centrifuge the original vial prior to removing the cap

Type: Primary
Antigen: Bone Morphogenetic Protein Receptor Type 1B
Clonality: monoclonal
Clone: 11C3
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR